siRNA Details
Gene Symbol: C1orf229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207401.2 | NP_997284.1 | 1 | 683 | ||
GenBank mRNA | AK127619.1 | - | 1 | 683 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
