siRNA Details
Gene Symbol: TUSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004125.2 | NP_001004125.1 | 1 | 746 | ||
GenBank mRNA | AY168647.2 | - | 1 | 328 | ||
BC017510.1 | - | 1 | 121 | |||
BC046919.1 | - | 1 | 187 | |||
BC110824.1 | - | 1 | 328 | 5169320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
