siRNA Details
Gene Symbol: C19orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317801.1 | NP_001304730.1 | 4 | 510 | ||
NM_033520.2 | NP_277055.1 | 4 | 451 | |||
GenBank mRNA | AB038317.1 | - | 4 | 448 | ||
AF213678.1 | - | 4 | 451 | |||
BC014264.2 | - | 4 | 408 | 4550805 | ||
BC060319.1 | - | 4 | 462 | 3901226 | ||
BX648823.1 | - | 4 | 427 | |||
CB109881.1 | - | 4 | 356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
