siRNA Details
Gene Symbol: SPANXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ238277.1 | - | 1, 2 | 103 | ||
AK225988.1 | - | 1, 2 | 122 | |||
BC054023.1 | - | 1, 2 | 125 | 6648369 |
Gene Symbol: SPANXC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA412605.1 | - | 1, 2 | 120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
