siRNA Details
Gene Symbol: SFTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205854.2 | NP_995326.1 | 1, 2 | 145 | ||
GenBank mRNA | AY102070.1 | - | 1, 2 | 120 | ||
AY359057.1 | - | 1, 2 | 123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
