siRNA Details
Gene Symbol: SFTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205854.2 | NP_995326.1 | 3 | 431 | ||
GenBank mRNA | AY102070.1 | - | 3 | 406 | ||
AY359057.1 | - | 3 | 409 | |||
BC039677.1 | - | 3 | 431 | 5186107 | ||
T94049.1 | - | 1 | 143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
