siRNA Details
Gene Symbol: WWP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270453.1 | NP_001257382.1 | 9 | 1084 | ||
NM_001270454.1 | NP_001257383.1 | 12 | 1394 | |||
NM_007014.4 | NP_008945.2 | 13 | 1447 | |||
NM_199424.2 | NP_955456.1 | 2 | 539 | |||
XM_011522823.2 | XP_011521125.1 | 14 | 1477 | |||
XM_011522825.1 | XP_011521127.1 | 12 | 1358 | |||
XM_011522826.2 | XP_011521128.1 | 9 | 1248 | |||
XM_017022879.1 | XP_016878368.1 | 14 | 1551 | |||
XM_017022880.1 | XP_016878369.1 | 14 | 1483 | |||
XM_017022881.1 | XP_016878370.1 | 14 | 1527 | |||
GenBank mRNA | AK126332.1 | - | 2 | 524 | ||
AK295070.1 | - | 5 | 439 | |||
AK295701.1 | - | 10 | 1143 | |||
AK300266.1 | - | 9 | 1084 | |||
AK312792.1 | - | 12 | 1374 | |||
AK316097.1 | - | 3 | 393 | |||
BC013645.1 | - | 11 | 1281 | 3453212 | ||
BC064531.1 | - | 12 | 1333 | 5588092 | ||
DB288556.1 | - | 2 | 539 | |||
JN712744.1 | - | 12 | 1364 | |||
U96114.1 | - | 13 | 1379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
