siRNA Details
Gene Symbol: WWP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270453.1 | NP_001257382.1 | 5 | 528 | ||
NM_001270454.1 | NP_001257383.1 | 8 | 838 | |||
NM_001270455.1 | NP_001257384.1 | 8 | 838 | |||
NM_007014.4 | NP_008945.2 | 9 | 891 | |||
XM_011522823.2 | XP_011521125.1 | 10 | 921 | |||
XM_011522825.1 | XP_011521127.1 | 8 | 802 | |||
XM_011522826.2 | XP_011521128.1 | 5 | 692 | |||
XM_017022879.1 | XP_016878368.1 | 10 | 995 | |||
XM_017022880.1 | XP_016878369.1 | 10 | 927 | |||
XM_017022881.1 | XP_016878370.1 | 10 | 971 | |||
GenBank mRNA | AJ001982.1 | - | 8 | 819 | ||
AK295701.1 | - | 6 | 587 | |||
AK300266.1 | - | 5 | 528 | |||
AK312792.1 | - | 8 | 818 | |||
BC000108.2 | - | 7 | 740 | |||
BC013645.1 | - | 7 | 725 | 3453212 | ||
BC064531.1 | - | 8 | 777 | 5588092 | ||
JN712744.1 | - | 8 | 808 | |||
U96114.1 | - | 9 | 823 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
