siRNA Details
Gene Symbol: WDFY3-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY250185.1 | - | 1 | 188 | ||
BC101193.2 | - | 1 | 188 | |||
BC101194.2 | - | 1 | 188 | |||
BC101195.1 | - | 1 | 188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
