siRNA Details
Gene Symbol: FRMD8P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033742.1 | 1 | 1435 | |||
GenBank mRNA | AF334945.1 | - | 1 | 1435 | ||
BC128150.1 | - | 1 | 1347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
