siRNA Details
Gene Symbol: KANK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198471.2 | NP_940873.2 | 2, 3 | 94 | ||
XM_006722718.3 | XP_006722781.1 | 2, 3 | 213 | |||
XM_011527884.2 | XP_011526186.1 | 2, 3 | 470 | |||
XM_017026563.1 | XP_016882052.1 | 1, 2 | 230 | |||
GenBank mRNA | AB284126.1 | - | 2, 3 | 94 | ||
AK128815.1 | - | 2, 3 | 94 | |||
BC035849.2 | - | 1, 2 | 69 | |||
BC066124.1 | - | 2, 3 | 94 | 30345663 | ||
BC066775.1 | - | 2, 3 | 94 | 30333203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
