siRNA Details
Gene Symbol: NPSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300933.1 | NP_001287862.1 | 4 | 577 | ||
NM_001300935.1 | NP_001287864.1 | 4 | 610 | |||
NM_207172.1 | NP_997055.1 | 4 | 564 | |||
NM_207173.1 | NP_997056.1 | 4 | 564 | |||
GenBank mRNA | AY255536.1 | - | 3 | 76 | ||
AY310326.1 | - | 4 | 564 | |||
AY310327.1 | - | 4 | 564 | |||
AY310328.1 | - | 4 | 531 | |||
AY310330.1 | - | 3 | 460 | |||
AY956446.1 | - | 4 | 469 | |||
AY956447.1 | - | 4 | 440 | |||
AY956448.1 | - | 4 | 440 | |||
BC132693.1 | - | 4 | 481 | 40146715 | ||
BC132695.1 | - | 4 | 481 | 40146717 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
