siRNA Details
Gene Symbol: MPDU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004870.3 | NP_004861.2 | 1 | 246 | ||
NR_024603.1 | 1 | 246 | ||||
XM_006721597.2 | XP_006721660.1 | 1 | 45 | |||
XM_006721598.2 | XP_006721661.1 | 1 | 45 | |||
XM_006721599.2 | XP_006721662.1 | 1 | 45 | |||
XM_011524081.2 | XP_011522383.1 | 1 | 41 | |||
GenBank mRNA | AF038961.1 | - | 1 | 46 | ||
AF059752.1 | - | 1 | 86 | |||
AK027742.1 | - | 1 | 45 | |||
AK055923.1 | - | 1 | 40 | |||
AK075299.1 | - | 1 | 45 | |||
AK225401.1 | - | 1 | 45 | |||
AK225432.1 | - | 1 | 41 | |||
AK297001.1 | - | 1 | 41 | |||
AK297691.1 | - | 1 | 41 | |||
AK300083.1 | - | 1 | 41 | |||
AK300140.1 | - | 1 | 37 | |||
AK314527.1 | - | 1 | 41 | |||
BC001898.1 | - | 1 | 57 | 3536216 | ||
BP325875.1 | - | 1 | 247 | |||
DC421138.1 | - | 1 | 247 | |||
DQ499597.1 | - | 1 | 30 | |||
KU178478.1 | - | 1 | 30 | |||
KU178479.1 | - | 1 | 30 | |||
KU178480.1 | - | 1 | 30 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
