siRNA Details
Gene Symbol: FAM3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171132.2 | NP_001164603.1 | 3 | 418 | ||
NM_001171134.2 | NP_001164605.1 | 2 | 515 | |||
NM_001282311.1 | NP_001269240.1 | 2 | 515 | |||
NM_001282312.1 | NP_001269241.1 | 3 | 359 | |||
NM_021806.3 | NP_068578.2 | 2 | 515 | |||
XM_005274714.3 | XP_005274771.1 | 3 | 365 | |||
XM_005274716.3 | XP_005274773.1 | 3 | 365 | |||
XM_005277879.4 | XP_005277936.1 | 3 | 424 | |||
XM_006724831.2 | XP_006724894.1 | 3 | 365 | |||
XM_006724832.3 | XP_006724895.1 | 3 | 424 | |||
XM_006724833.2 | XP_006724896.1 | 3 | 365 | |||
XM_006724834.2 | XP_006724897.1 | 2 | 423 | |||
XM_017029699.1 | XP_016885188.1 | 2 | 522 | |||
XM_017029700.1 | XP_016885189.1 | 2 | 521 | |||
XM_017029701.1 | XP_016885190.1 | 3 | 424 | |||
GenBank mRNA | AK024601.1 | - | 3 | 375 | ||
AK057945.1 | - | 3 | 233 | |||
AK294116.1 | - | 2 | 478 | |||
AK314682.1 | - | 2 | 433 | |||
AL539139.3 | - | 2 | 512 | |||
BC002934.1 | - | 2 | 419 | |||
BC008912.2 | - | 2 | 166 | 3639043 | ||
BM457676.1 | - | 3 | 400 | |||
X87193.1 | - | 2 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
