siRNA Details
Gene Symbol: ZXDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007156.4 | NP_009087.1 | 1 | 435 | ||
GenBank mRNA | AK122815.1 | - | 1 | 435 | ||
AK292686.1 | - | 1 | 425 | |||
BC059356.1 | - | 1 | 233 | 30347480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
