siRNA Details
Gene Symbol: PLEKHG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129727.2 | NP_001123199.1 | 3 | 757 | ||
NM_001129728.1 | NP_001123200.1 | 2 | 717 | |||
NM_001129729.2 | NP_001123201.1 | 2 | 690 | |||
XM_011522985.2 | XP_011521287.1 | 2 | 687 | |||
XM_011522986.2 | XP_011521288.1 | 2 | 819 | |||
XM_011522987.2 | XP_011521289.1 | 3 | 763 | |||
XM_011522988.2 | XP_011521290.1 | 2 | 636 | |||
GenBank mRNA | AB197663.1 | - | 1 | 2953 | ||
AB197664.1 | - | 1 | 2953 | |||
AK024475.1 | - | 2 | 652 | |||
AK308298.1 | - | 2 | 626 | |||
BC063501.1 | - | 2 | 628 | 5527705 | ||
BC082974.1 | - | 3 | 725 | 6291175 | ||
BC115390.1 | - | 1 | 567 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
