siRNA Details
Gene Symbol: PCAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026555.1 | 1 | 57 | |||
GenBank mRNA | AF388200.1 | - | 1 | 52 | ||
AK056825.1 | - | 1 | 57 | |||
BC105591.1 | - | 1 | 155 | |||
BC125139.1 | - | 1 | 54 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
