siRNA Details
Gene Symbol: C5orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017987.2 | NP_001017987.1 | 4 | 372 | ||
NM_016175.3 | NP_057259.2 | 6 | 537 | |||
GenBank mRNA | AF153685.1 | - | 2 | 158 | ||
AK293901.1 | - | 6 | 503 | |||
AK295104.1 | - | 4 | 365 | |||
AK298954.1 | - | 4 | 329 | |||
BC002739.2 | - | 2 | 84 | |||
BC056889.1 | - | 5 | 401 | |||
BC069051.1 | - | 6 | 505 | 3945859 | ||
BF965313.1 | - | 5 | 461 | |||
BQ642639.1 | - | 4 | 328 | |||
BX537968.1 | - | 7 | 638 | |||
BX647375.1 | - | 6 | 505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
