siRNA Details
Gene Symbol: C20orf203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182584.2 | NP_872390.2 | 3 | 538 | ||
GenBank mRNA | AK091025.1 | - | 3 | 435 | ||
BC105014.1 | - | 3 | 314 | |||
BC105042.1 | - | 3 | 316 | |||
GU931819.1 | - | 4 | 848 | |||
GU931820.1 | - | 4 | 848 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
