siRNA Details
Gene Symbol: LRRC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201550.3 | NP_963844.2 | 1 | 772 | ||
GenBank mRNA | AK123908.1 | - | 1 | 368 | ||
BC090881.1 | - | 1 | 772 | 30530394 | ||
BC106892.1 | - | 1 | 365 | |||
BC106893.1 | - | 1 | 368 | 40030470 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
