siRNA Details
Gene Symbol: ROPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317774.1 | NP_001304703.1 | 2 | 337 | ||
NM_017578.3 | NP_060048.2 | 3 | 417 | |||
NR_133916.1 | 2 | 337 | ||||
NR_133917.1 | 3 | 261 | ||||
NR_133918.1 | 3 | 494 | ||||
NR_133919.1 | 2 | 448 | ||||
XM_011512933.2 | XP_011511235.1 | 3 | 381 | |||
XM_011512934.2 | XP_011511236.1 | 3 | 492 | |||
GenBank mRNA | AF303889.2 | - | 3 | 417 | ||
AK310283.1 | - | 2 | 387 | |||
AL133624.1 | - | 2 | 260 | |||
BC067767.1 | - | 2 | 304 | |||
BC111005.1 | - | 3 | 178 | |||
BC132744.1 | - | 1 | 173 | 40146766 | ||
BC132746.1 | - | 1 | 174 | 40146768 | ||
BG724071.1 | - | 2 | 305 | |||
DB042884.1 | - | 3 | 465 | |||
HY055534.1 | - | 2 | 337 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
