siRNA Details
Gene Symbol: NANS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018946.3 | NP_061819.2 | 5 | 936 | ||
XM_011518787.2 | XP_011517089.1 | 5 | 692 | |||
XM_011518788.2 | XP_011517090.1 | 3 | 443 | |||
XM_017014811.1 | XP_016870300.1 | 4 | 637 | |||
GenBank mRNA | AF161387.1 | - | 2 | 234 | ||
AF257466.1 | - | 5 | 804 | |||
AK001659.1 | - | 5 | 874 | |||
AK091184.1 | - | 3 | 2730 | |||
AK316608.1 | - | 5 | 896 | |||
AL526281.3 | - | 5 | 939 | |||
BC000008.2 | - | 5 | 882 | 3505324 | ||
BC019315.1 | - | 5 | 864 | 2821389 | ||
FJ224349.1 | - | 5 | 804 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
