siRNA Details
Gene Symbol: C20orf27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039140.2 | NP_001034229.1 | 6 | 792 | ||
NM_001258429.1 | NP_001245358.1 | 6 | 717 | |||
NM_001258430.1 | NP_001245359.1 | 6 | 1171 | |||
NR_047675.1 | 6 | 861 | ||||
XM_011529266.2 | XP_011527568.1 | 5 | 649 | |||
GenBank mRNA | AK000557.1 | - | 6 | 656 | ||
AK024220.1 | - | 6 | 656 | |||
AK097645.1 | - | 6 | 1299 | |||
AK124341.1 | - | 4 | 1909 | |||
AK125202.1 | - | 3 | 1863 | |||
AK225109.1 | - | 6 | 656 | |||
AK290955.1 | - | 6 | 861 | |||
BC012196.1 | - | 6 | 643 | 2822218 | ||
BC024036.1 | - | 6 | 654 | 3510659 | ||
BG474804.1 | - | 6 | 713 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
