siRNA Details
Gene Symbol: C20orf27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039140.2 | NP_001034229.1 | 5, 6 | 738 | ||
NM_001258429.1 | NP_001245358.1 | 5, 6 | 663 | |||
NM_001258430.1 | NP_001245359.1 | 5, 6 | 1117 | |||
NR_047675.1 | 5, 6 | 807 | ||||
XM_011529266.2 | XP_011527568.1 | 4, 5 | 595 | |||
GenBank mRNA | AK000557.1 | - | 5, 6 | 602 | ||
AK024220.1 | - | 5, 6 | 602 | |||
AK097645.1 | - | 5, 6 | 1245 | |||
AK124341.1 | - | 4 | 1855 | |||
AK125202.1 | - | 3 | 1809 | |||
AK225109.1 | - | 5, 6 | 602 | |||
AK290955.1 | - | 5, 6 | 807 | |||
BC012196.1 | - | 5, 6 | 589 | 2822218 | ||
BC024036.1 | - | 5, 6 | 600 | 3510659 | ||
BG474804.1 | - | 5, 6 | 658 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
