siRNA Details
Gene Symbol: CCDC130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320561.1 | NP_001307490.1 | 11 | 2025 | ||
NM_001320564.1 | NP_001307493.1 | 11 | 2013 | |||
NM_001320565.1 | NP_001307494.1 | 11 | 1895 | |||
NM_001320566.1 | NP_001307495.1 | 10 | 2102 | |||
NM_001320567.1 | NP_001307496.1 | 10 | 1965 | |||
NM_001320568.1 | NP_001307497.1 | 9 | 1774 | |||
NM_001320569.1 | NP_001307498.1 | 9 | 1481 | |||
NM_030818.3 | NP_110445.1 | 10 | 1685 | |||
XM_005260086.4 | XP_005260143.1 | 11 | 1875 | |||
GenBank mRNA | AA480850.1 | - | 1 | 264 | ||
AF250306.1 | - | 10 | 1613 | |||
AK098262.1 | - | 10 | 1642 | |||
BC002905.2 | - | 10 | 1609 | 3940130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
