siRNA Details
Gene Symbol: CCDC130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320561.1 | NP_001307490.1 | 11 | 1384 | ||
NM_001320564.1 | NP_001307493.1 | 11 | 1372 | |||
NM_001320565.1 | NP_001307494.1 | 11 | 1254 | |||
NM_001320566.1 | NP_001307495.1 | 10 | 1461 | |||
NM_001320567.1 | NP_001307496.1 | 10 | 1324 | |||
NM_001320568.1 | NP_001307497.1 | 9 | 1133 | |||
NM_001320569.1 | NP_001307498.1 | 9 | 840 | |||
NM_030818.3 | NP_110445.1 | 10 | 1044 | |||
XM_005260086.4 | XP_005260143.1 | 11 | 1234 | |||
GenBank mRNA | AF250306.1 | - | 10 | 972 | ||
AK098262.1 | - | 10 | 1001 | |||
BC002905.2 | - | 10 | 968 | 3940130 | ||
BI913255.1 | - | 9 | 808 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
