siRNA Details
Gene Symbol: KAZALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319303.1 | NP_001306232.1 | 6 | 2639 | ||
NR_135067.1 | 5 | 2022 | ||||
NR_135068.1 | 2 | 971 | ||||
XM_017016714.1 | XP_016872203.1 | 5 | 1663 | |||
XM_017016715.1 | XP_016872204.1 | 5 | 1640 | |||
GenBank mRNA | AF333487.1 | - | 3 | 764 | ||
AK172864.1 | - | 7 | 2550 | |||
AY014271.1 | - | 3 | 764 | |||
BC012394.1 | - | 2 | 871 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
