siRNA Details
Gene Symbol: KAZALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319303.1 | NP_001306232.1 | 2 | 1108 | ||
NM_030929.4 | NP_112191.2 | 2 | 1108 | |||
XM_005270194.3 | XP_005270251.1 | 2 | 1209 | |||
XM_017016714.1 | XP_016872203.1 | 1 | 187 | |||
XM_017016715.1 | XP_016872204.1 | 1 | 187 | |||
GenBank mRNA | AF333487.1 | - | 1 | 273 | ||
AK172862.1 | - | 2 | 359 | |||
AK172864.1 | - | 2 | 1107 | |||
AK225949.1 | - | 2 | 360 | |||
AY014271.1 | - | 1 | 273 | |||
AY359087.1 | - | 2 | 364 | |||
BC007758.2 | - | 2 | 509 | 4122366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
