siRNA Details
Gene Symbol: RUNX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031680.2 | NP_001026850.1 | 2 | 518 | ||
NM_001320672.1 | NP_001307601.1 | 3 | 351 | |||
NM_004350.2 | NP_004341.1 | 1 | 448 | |||
XM_005246024.4 | XP_005246081.1 | 3 | 226 | |||
XM_011542351.1 | XP_011540653.1 | 2 | 518 | |||
XM_017002670.1 | XP_016858159.1 | 3 | 226 | |||
GenBank mRNA | AF022726.2 | - | 3 | 189 | ||
AK301044.1 | - | 2 | 121 | |||
AK310945.1 | - | 2 | 514 | |||
BC013362.2 | - | 3 | 325 | 3615873 | ||
DA935726.1 | - | 2 | 518 | |||
X79550.2 | - | 1 | 43 | |||
Z35278.1 | - | 1 | 46 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
