siRNA Details
Gene Symbol: RUNX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031680.2 | NP_001026850.1 | 2 | 572 | ||
NM_001320672.1 | NP_001307601.1 | 3 | 405 | |||
NM_004350.2 | NP_004341.1 | 1 | 502 | |||
XM_005246024.4 | XP_005246081.1 | 3 | 280 | |||
XM_011542351.1 | XP_011540653.1 | 2 | 572 | |||
XM_017002670.1 | XP_016858159.1 | 3 | 280 | |||
GenBank mRNA | AF022726.2 | - | 3 | 243 | ||
AK301044.1 | - | 2 | 175 | |||
AK310945.1 | - | 2 | 568 | |||
BC013362.2 | - | 3 | 379 | 3615873 | ||
DA935726.1 | - | 2 | 572 | |||
X79550.2 | - | 1 | 97 | |||
Z35278.1 | - | 1 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
