siRNA Details
Gene Symbol: TBC1D20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144628.3 | NP_653229.1 | 5 | 755 | ||
NR_111901.1 | 5 | 755 | ||||
XM_005260661.1 | XP_005260718.1 | 5 | 745 | |||
XM_006723540.3 | XP_006723603.1 | 4 | 534 | |||
XM_017027645.1 | XP_016883134.1 | 4 | 526 | |||
GenBank mRNA | AB449906.1 | - | 5 | 607 | ||
AK055573.1 | - | 1 | 36 | |||
AK095110.1 | - | 4 | 914 | |||
AK127062.1 | - | 5 | 735 | |||
AK291648.1 | - | 5 | 718 | |||
BC014983.1 | - | 5 | 705 | 4861869 | ||
BX647370.1 | - | 1 | 286 | |||
DA230581.1 | - | 2 | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
