siRNA Details
Gene Symbol: TBC1D20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144628.3 | NP_653229.1 | 8 | 3157 | ||
XM_005260661.1 | XP_005260718.1 | 9 | 2196 | |||
XM_006723540.3 | XP_006723603.1 | 7 | 2936 | |||
XM_017027645.1 | XP_016883134.1 | 8 | 1977 | |||
GenBank mRNA | AK092063.1 | - | 1 | 943 | ||
AK095110.1 | - | 7 | 3316 | |||
AK127062.1 | - | 8 | 3138 | |||
BX647370.1 | - | 3 | 3888 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
