siRNA Details
Gene Symbol: SGSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301849.1 | NP_001288778.1 | 14 | 1692 | ||
NM_015705.5 | NP_056520.2 | 15 | 1837 | |||
XM_005261572.2 | XP_005261629.1 | 15 | 1864 | |||
XM_005261573.3 | XP_005261630.1 | 15 | 1869 | |||
XM_005261574.4 | XP_005261631.1 | 15 | 1869 | |||
XM_005261575.1 | XP_005261632.1 | 14 | 1732 | |||
XM_011530158.1 | XP_011528460.1 | 14 | 1688 | |||
XM_011530159.1 | XP_011528461.1 | 15 | 1752 | |||
XM_011530160.1 | XP_011528462.1 | 14 | 1669 | |||
XM_011530161.1 | XP_011528463.1 | 15 | 1861 | |||
XM_011530162.2 | XP_011528464.1 | 15 | 1747 | |||
XM_017028775.1 | XP_016884264.1 | 14 | 1656 | |||
XM_017028776.1 | XP_016884265.1 | 14 | 1725 | |||
XM_017028777.1 | XP_016884266.1 | 14 | 1725 | |||
XM_017028778.1 | XP_016884267.1 | 13 | 1588 | |||
XM_017028779.1 | XP_016884268.1 | 14 | 1778 | |||
XM_017028780.1 | XP_016884269.1 | 14 | 1767 | |||
GenBank mRNA | AB196956.1 | - | 13 | 1606 | ||
AB275763.1 | - | 14 | 1710 | |||
AB449880.1 | - | 13 | 1599 | |||
AK000006.1 | - | 6 | 2591 | |||
AK295469.1 | - | 15 | 1783 | |||
AK297599.1 | - | 14 | 1685 | |||
AY308849.1 | - | 15 | 1772 | |||
BC008078.2 | - | 15 | 1763 | 3027777 | ||
BC127850.1 | - | 8 | 913 | |||
CR456441.1 | - | 14 | 1635 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
