siRNA Details
Gene Symbol: SGSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301849.1 | NP_001288778.1 | 21 | 2382 | ||
NM_015705.5 | NP_056520.2 | 22 | 2605 | |||
XM_005261572.2 | XP_005261629.1 | 22 | 2683 | |||
XM_005261575.1 | XP_005261632.1 | 21 | 2551 | |||
XM_011530158.1 | XP_011528460.1 | 21 | 2507 | |||
XM_011530159.1 | XP_011528461.1 | 22 | 2571 | |||
XM_011530160.1 | XP_011528462.1 | 21 | 2488 | |||
XM_011530161.1 | XP_011528463.1 | 22 | 2680 | |||
XM_011530162.2 | XP_011528464.1 | 22 | 2515 | |||
XM_017028775.1 | XP_016884264.1 | 21 | 2424 | |||
XM_017028776.1 | XP_016884265.1 | 21 | 2493 | |||
XM_017028778.1 | XP_016884267.1 | 20 | 2407 | |||
XM_017028779.1 | XP_016884268.1 | 21 | 2597 | |||
XM_017028780.1 | XP_016884269.1 | 21 | 2535 | |||
GenBank mRNA | AB196956.1 | - | 20 | 2374 | ||
AB275763.1 | - | 21 | 2478 | |||
AK000006.1 | - | 9 | 3992 | |||
AK301042.1 | - | 21 | 2295 | |||
AL137499.1 | - | 5 | 525 | |||
AY308849.1 | - | 22 | 2540 | |||
BC008078.2 | - | 22 | 2531 | 3027777 | ||
BG741967.1 | - | 40 | ||||
CR456441.1 | - | 21 | 2403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
