siRNA Details
Gene Symbol: CCL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002984.3 | NP_002975.1 | 3 | 295 | ||
GenBank mRNA | AY312578.1 | - | 2 | 112 | ||
AY766446.1 | - | 3 | 295 | |||
AY766448.1 | - | 2 | 180 | |||
BC104226.1 | - | 3 | 286 | 40032168 | ||
BC104227.1 | - | 3 | 286 | 40032172 | ||
BC107433.1 | - | 3 | 296 | 6727378 | ||
CR542119.1 | - | 3 | 216 | |||
J04130.1 | - | 3 | 324 | |||
M23502.1 | - | 3 | 290 | |||
M25316.1 | - | 3 | 289 | |||
M57503.1 | - | 3 | 203 | |||
X16166.1 | - | 3 | 268 | |||
X53683.1 | - | 3 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
