siRNA Details
Gene Symbol: KANK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320269.1 | NP_001307198.1 | 9 | 2446 | ||
NM_181712.4 | NP_859063.3 | 10 | 4328 | |||
XM_011540843.2 | XP_011539145.1 | 10 | 4213 | |||
XM_011540845.2 | XP_011539147.1 | 10 | 4207 | |||
XM_017000483.1 | XP_016855972.1 | 9 | 4077 | |||
XM_017000484.1 | XP_016855973.1 | 10 | 2582 | |||
XM_017000485.1 | XP_016855974.1 | 8 | 2514 | |||
GenBank mRNA | AB284127.1 | - | 10 | 4328 | ||
AK096259.1 | - | 4 | 1712 | |||
AL832644.2 | - | 8 | 3080 | |||
BC060866.1 | - | 10 | 4328 | 30344114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
