siRNA Details
Gene Symbol: ASUN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018164.2 | NP_060634.2 | 14 | 2126 | ||
XM_005253422.3 | XP_005253479.1 | 13 | 1914 | |||
XM_011520750.2 | XP_011519052.1 | 14 | 1677 | |||
XM_011520751.2 | XP_011519053.1 | 13 | 1932 | |||
XM_011520752.2 | XP_011519054.1 | 13 | 1878 | |||
XM_011520753.2 | XP_011519055.1 | 8 | 849 | |||
XM_017019632.1 | XP_016875121.1 | 14 | 1659 | |||
XM_017019633.1 | XP_016875122.1 | 13 | 2069 | |||
XM_017019634.1 | XP_016875123.1 | 13 | 2051 | |||
XM_017019635.1 | XP_016875124.1 | 13 | 1488 | |||
XM_017019636.1 | XP_016875125.1 | 13 | 1860 | |||
XM_017019637.1 | XP_016875126.1 | 12 | 1289 | |||
GenBank mRNA | AF274950.1 | - | 5 | 538 | ||
AK001222.1 | - | 4 | 441 | |||
AK001492.1 | - | 14 | 1755 | |||
AK001499.1 | - | 14 | 1791 | |||
AK225336.1 | - | 14 | 1777 | |||
AK297948.1 | - | 12 | 1466 | |||
BC003081.2 | - | 14 | 1815 | 2989678 | ||
BC008368.1 | - | 14 | 1773 | 3355282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
