siRNA Details
Gene Symbol: ASUN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018164.2 | NP_060634.2 | 14 | 2150 | ||
XM_005253422.3 | XP_005253479.1 | 13 | 1938 | |||
XM_011520750.2 | XP_011519052.1 | 14 | 1701 | |||
XM_011520751.2 | XP_011519053.1 | 13 | 1956 | |||
XM_011520752.2 | XP_011519054.1 | 13 | 1902 | |||
XM_011520753.2 | XP_011519055.1 | 8 | 873 | |||
XM_017019632.1 | XP_016875121.1 | 14 | 1683 | |||
XM_017019633.1 | XP_016875122.1 | 13 | 2093 | |||
XM_017019634.1 | XP_016875123.1 | 13 | 2075 | |||
XM_017019635.1 | XP_016875124.1 | 13 | 1512 | |||
XM_017019636.1 | XP_016875125.1 | 13 | 1884 | |||
XM_017019637.1 | XP_016875126.1 | 12 | 1313 | |||
GenBank mRNA | AF274950.1 | - | 5 | 562 | ||
AK001222.1 | - | 4 | 465 | |||
AK001499.1 | - | 14 | 1815 | |||
AK225336.1 | - | 14 | 1801 | |||
AK297948.1 | - | 12 | 1490 | |||
BC003081.2 | - | 14 | 1839 | 2989678 | ||
BC008368.1 | - | 14 | 1797 | 3355282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
