siRNA Details
Gene Symbol: FOXN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213596.2 | NP_998761.2 | 9 | 1340 | ||
XM_011537922.2 | XP_011536224.1 | 9 | 1529 | |||
XM_017018818.1 | XP_016874307.1 | 7 | 2315 | |||
GenBank mRNA | AF425596.1 | - | 7 | 1115 | ||
AF425597.1 | - | 6 | 990 | |||
AK131519.1 | - | 5 | 947 | |||
BC113073.1 | - | 2 | 340 | |||
BC146825.1 | - | 9 | 1341 | 8860453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
