siRNA Details
Gene Symbol: FOXN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213596.2 | NP_998761.2 | 8 | 962 | ||
XM_011537922.2 | XP_011536224.1 | 8 | 1151 | |||
XM_011537923.2 | XP_011536225.1 | 8 | 1151 | |||
XM_017018818.1 | XP_016874307.1 | 6 | 1937 | |||
XM_017018819.1 | XP_016874308.1 | 8 | 1151 | |||
GenBank mRNA | AF425596.1 | - | 6 | 737 | ||
AF425597.1 | - | 5 | 612 | |||
AK131519.1 | - | 4 | 569 | |||
BC146825.1 | - | 8 | 963 | 8860453 |

More Information
Other Species Targeted

Gene Ontology Categories:
