siRNA Details
Gene Symbol: PIGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033198.3 | NP_149975.1 | 12 | 2659 | ||
GenBank mRNA | AF161427.1 | - | 6 | 1522 | ||
AK025825.1 | - | 5 | 1426 | |||
AK055512.1 | - | 7 | 1491 | |||
AK075428.1 | - | 12 | 2316 | |||
AK127162.1 | - | 3 | 2231 | |||
AY359112.1 | - | 11 | 2556 | |||
BC001319.2 | - | 4 | 1358 | |||
BC007301.1 | - | 10 | 2045 | |||
BC069228.1 | - | 12 | 2311 | 3944034 | ||
BX640839.1 | - | 2 | 2873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
