siRNA Details
Gene Symbol: GON4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282856.1 | NP_001269785.1 | 21 | 3673 | ||
NM_001282858.1 | NP_001269787.1 | 21 | 3725 | |||
NM_001282860.1 | NP_001269789.1 | 21 | 3725 | |||
NM_001282861.1 | NP_001269790.1 | 21 | 3725 | |||
NM_032292.5 | NP_115668.4 | 21 | 3713 | |||
XM_005245284.3 | XP_005245341.1 | 21 | 3665 | |||
XM_005245286.3 | XP_005245343.1 | 6 | 1476 | |||
XM_006711393.3 | XP_006711456.1 | 21 | 3684 | |||
XM_006711394.3 | XP_006711457.1 | 21 | 3670 | |||
XM_011509658.2 | XP_011507960.1 | 20 | 3565 | |||
XM_011509659.2 | XP_011507961.1 | 21 | 3689 | |||
GenBank mRNA | AB046826.1 | - | 1 | 116 | ||
AB195687.1 | - | 21 | 3725 | |||
AB232668.1 | - | 21 | 3601 | |||
AK022985.1 | - | 5 | 1237 | |||
AK222851.1 | - | 7 | 1519 | |||
AK315191.1 | - | 11 | 1948 | |||
AL136565.1 | - | 7 | 1581 | |||
AY335490.2 | - | 21 | 3628 | |||
AY335491.2 | - | 21 | 3673 | |||
AY335492.2 | - | 21 | 3599 | |||
BC036740.2 | - | 10 | 1906 | |||
BC114629.1 | - | 21 | 3600 | 40082464 | ||
BC115703.1 | - | 21 | 3612 | 40082468 | ||
BC117557.1 | - | 6 | 1488 | |||
BX537764.1 | - | 21 | 3601 | |||
BX648043.1 | - | 21 | 3624 | |||
BX648802.1 | - | 20 | 3958 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
