siRNA Details
Gene Symbol: GON4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282856.1 | NP_001269785.1 | 21 | 4362 | ||
NM_001282858.1 | NP_001269787.1 | 21 | 4414 | |||
NM_001282860.1 | NP_001269789.1 | 21 | 4414 | |||
NM_001282861.1 | NP_001269790.1 | 21 | 4414 | |||
NM_032292.5 | NP_115668.4 | 21 | 4402 | |||
XM_005245284.3 | XP_005245341.1 | 21 | 4354 | |||
XM_005245286.3 | XP_005245343.1 | 6 | 2165 | |||
XM_006711393.3 | XP_006711456.1 | 21 | 4373 | |||
XM_006711394.3 | XP_006711457.1 | 21 | 4359 | |||
XM_011509658.2 | XP_011507960.1 | 20 | 4254 | |||
XM_011509659.2 | XP_011507961.1 | 21 | 4378 | |||
GenBank mRNA | AB046826.1 | - | 1 | 805 | ||
AB195687.1 | - | 21 | 4414 | |||
AB195688.1 | - | 22 | 3096 | |||
AB232668.1 | - | 21 | 4290 | |||
AK022985.1 | - | 5 | 1926 | |||
AK222851.1 | - | 7 | 2208 | |||
AK315191.1 | - | 11 | 2637 | |||
AL136565.1 | - | 7 | 2270 | |||
AY335490.2 | - | 21 | 4317 | |||
AY335491.2 | - | 21 | 4362 | |||
AY335492.2 | - | 21 | 4288 | |||
BC036740.2 | - | 10 | 2595 | |||
BC114629.1 | - | 21 | 4289 | 40082464 | ||
BC115703.1 | - | 21 | 4301 | 40082468 | ||
BC117557.1 | - | 6 | 2177 | |||
BX537764.1 | - | 21 | 4290 | |||
BX648043.1 | - | 21 | 4313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
