siRNA Details
Gene Symbol: FBXL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321213.1 | NP_001308142.1 | 3 | 1860 | ||
NM_024963.5 | NP_079239.3 | 3 | 1860 | |||
XM_011515531.2 | XP_011513833.1 | 3 | 1875 | |||
XM_011515532.2 | XP_011513834.1 | 3 | 1875 | |||
GenBank mRNA | AK021529.1 | - | 1 | 467 | ||
AK057042.1 | - | 3 | 1814 | |||
AK095394.1 | - | 1 | 1387 | |||
AK294949.1 | - | 3 | 1854 | |||
BC004228.1 | - | 1 | 313 | |||
BC013435.1 | - | 1 | 462 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
