siRNA Details
Gene Symbol: YTHDF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017798.3 | NP_060268.2 | 4 | 1345 | ||
GenBank mRNA | AB055518.1 | - | 1 | 686 | ||
AK000398.1 | - | 2 | 505 | |||
AK095795.1 | - | 4 | 1063 | |||
AK300073.1 | - | 2 | 1075 | |||
AL834366.1 | - | 1 | 466 | |||
BC003681.1 | - | 3 | 1019 | |||
BC016920.2 | - | 1 | 762 | |||
BC025264.1 | - | 1 | 882 | |||
BC050284.1 | - | 4 | 1345 | 5298438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
