siRNA Details
Gene Symbol: PPP1R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002711.3 | NP_002702.2 | 3, 4 | 997 | ||
XM_005250473.3 | XP_005250530.1 | 4, 5 | 543 | |||
GenBank mRNA | AF024579.1 | - | 4, 5 | 445 | ||
BC126451.1 | - | 3, 4 | 1035 | 8992167 | ||
BC126453.1 | - | 3, 4 | 1035 | 8992169 | ||
X78578.1 | - | 3, 4 | 964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
