siRNA Details
Gene Symbol: ALS2CR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168216.1 | NP_001161688.1 | 13 | 1505 | ||
NM_001168221.1 | NP_001161693.1 | 16 | 5440 | |||
NM_152525.5 | NP_689738.3 | 15 | 1849 | |||
XM_006712331.3 | XP_006712394.1 | 15 | 1852 | |||
XM_006712332.3 | XP_006712395.1 | 14 | 1777 | |||
GenBank mRNA | AB053313.1 | - | 2 | 1197 | ||
AK058080.1 | - | 13 | 1477 | |||
AK097651.1 | - | 2 | 2455 | |||
BC030659.2 | - | 15 | 1842 | 4838801 | ||
BX648732.1 | - | 16 | 5433 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
