siRNA Details
Gene Symbol: ZNF667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321355.1 | NP_001308284.1 | 6 | 698 | ||
NM_001321356.1 | NP_001308285.1 | 7 | 1199 | |||
NM_022103.3 | NP_071386.3 | 5 | 597 | |||
XM_011527208.1 | XP_011525510.1 | 6 | 2843 | |||
XM_011527209.1 | XP_011525511.1 | 5 | 2357 | |||
GenBank mRNA | AK096513.1 | - | 7 | 1128 | ||
BC150593.1 | - | 4 | 433 | 9056963 | ||
CR936605.1 | - | 6 | 663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
