siRNA Details
Gene Symbol: U2AF1L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040425.2 | NP_001035515.1 | 4 | 337 | ||
NM_144987.3 | NP_659424.2 | 4 | 337 | |||
NR_110171.1 | 5 | 425 | ||||
NR_110172.1 | 4 | 602 | ||||
NR_110173.1 | 5 | 450 | ||||
GenBank mRNA | AK092844.1 | - | 2 | 366 | ||
AK094213.1 | - | 2 | 1189 | |||
AK303983.1 | - | 3 | 1015 | |||
AY461582.1 | - | 5 | 399 | |||
AY569437.3 | - | 4 | 306 | |||
BC021186.1 | - | 4 | 306 | 5260387 | ||
BM696851.1 | - | 5 | 443 | |||
CD175846.1 | - | 4 | 346 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_170760, BC060972, AF419339.
Gene Ontology Categories:
