siRNA Details
Gene Symbol: U2AF1L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144987.3 | NP_659424.2 | 4, 5 | 365 | ||
NR_110172.1 | 4, 5 | 630 | ||||
NR_110173.1 | 5, 6 | 478 | ||||
GenBank mRNA | AK092844.1 | - | 2, 3 | 394 | ||
AK094213.1 | - | 2, 3 | 1217 | |||
AK303983.1 | - | 3, 4 | 1043 | |||
BC021186.1 | - | 4, 5 | 334 | 5260387 | ||
BM696851.1 | - | 5, 6 | 471 | |||
CD175846.1 | - | 4, 5 | 374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
